Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leriglitazone (hydrochloride) | 146062-46-6 | 99.6% | 408.90 g·mol⁻¹ | C19H21ClN2O4S | 25 MG
SDP

Catalog No. 5000228223
Encompass_Preferred

Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone, an orally bioavailable, blood-brain-barrier-penetrant PPARγ agonist supplied for research use. The compound is provided as a 25 mg solid vial with supporting product and safety documentation available from the supplier.

  • High reported purity (99.64%).
  • Molecular weight 408.90 g·mol⁻¹.
  • Chemical formula C19H21ClN2O4S.
  • Supplied as a 25 mg solid vial and 10 mM DMSO stock options.
  • Recommended storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • Product datasheet and SDS provided for handling and safety guidance.

Catalog No. 50-002-28223 Supplier Medchemexpress LLC Supplier No. HY117727A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone, an orally bioavailable, blood-brain-barrier-penetrant PPARγ agonist supplied for research use. The compound is provided as a 25 mg solid vial with supporting product and safety documentation available from the supplier.

  • High reported purity (99.64%).
  • Molecular weight 408.90 g·mol⁻¹.
  • Chemical formula C19H21ClN2O4S.
  • Supplied as a 25 mg solid vial and 10 mM DMSO stock options.
  • Recommended storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • Product datasheet and SDS provided for handling and safety guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.