Learn More
Medchemexpress LLC Leriglitazone hydrochloride | 146062-46-6 | MFCD00055081 | 99.6% | 408.90 | C19H21ClN2O4S | 1 ML
SDP
Catalog No. 5000226275
Encompass_Preferred
Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone (MIN-102), a brain-penetrant PPARγ agonist used in preclinical research on neuroinflammation, mitochondrial function, and neurodegeneration. Supplied for research use as a 10 mM solution in DMSO (1 mL) and as solid quantities for custom preparation.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for cell-based or biochemical assays.
  • High purity (≈99.6%) suitable for research applications.
  • Also available as solid material for custom dilution and dosing.
  • Useful for studies of PPARγ-mediated neuroprotection and mitochondrial regulation.
  • Provided with supporting datasheet and SDS for handling and safety.
Catalog No. 50-002-26275 Supplier Medchemexpress LLC Supplier No. HY117727A10MM 1ML
Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone (MIN-102), a brain-penetrant PPARγ agonist used in preclinical research on neuroinflammation, mitochondrial function, and neurodegeneration. Supplied for research use as a 10 mM solution in DMSO (1 mL) and as solid quantities for custom preparation.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for cell-based or biochemical assays.
  • High purity (≈99.6%) suitable for research applications.
  • Also available as solid material for custom dilution and dosing.
  • Useful for studies of PPARγ-mediated neuroprotection and mitochondrial regulation.
  • Provided with supporting datasheet and SDS for handling and safety.