Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leriglitazone (hydrochloride) | 146062-46-6 | MFCD21734117 | 99.6% | 408.90 g·mol⁻¹ | C19H21ClN2O4S | 100 MG
SDP

Catalog No. 5000228783
Encompass_Preferred

Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone, a blood-brain barrier-penetrant PPARγ agonist used in preclinical research on neuroinflammation and neurodegeneration. Supplied as a solid for laboratory research use only.

  • High purity, typically 99.6% by HPLC.
  • Molecular weight 408.90 g·mol⁻¹ and formula C19H21ClN2O4S.
  • Available in multiple solid sizes, including 100 MG, and as DMSO solutions.
  • Suitable for in vitro and in vivo preclinical studies focused on PPARγ modulation.
  • Provided with an SDS and product data sheet for safe handling and characterization.

Catalog No. 50-002-28783 Supplier Medchemexpress LLC Supplier No. HY117727A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone, a blood-brain barrier-penetrant PPARγ agonist used in preclinical research on neuroinflammation and neurodegeneration. Supplied as a solid for laboratory research use only.

  • High purity, typically 99.6% by HPLC.
  • Molecular weight 408.90 g·mol⁻¹ and formula C19H21ClN2O4S.
  • Available in multiple solid sizes, including 100 MG, and as DMSO solutions.
  • Suitable for in vitro and in vivo preclinical studies focused on PPARγ modulation.
  • Provided with an SDS and product data sheet for safe handling and characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.