Learn More
Medchemexpress LLC Leritrelvir (RAY1216) | 2923310-64-7 | C31H44F3N5O6 | 5 MG
SDP
Catalog No. 5000373573
Encompass_Preferred
Leritrelvir (RAY1216) is an orally active SARS-CoV-2 main protease slow-tight inhibitor with a Ki of 8.6 nM. It is for research use only and not sold to patients.
  • Orally active SARS-CoV-2 main protease slow-tight inhibitor
  • Potent inhibitor with a Ki of 8.6 nM
  • Covalently attaches to the catalytic Cys145
  • Demonstrates antiviral activities against SARS-CoV-2 wild type ancestral strain and variants
  • Effectively prolongs survival in SARS-CoV-2 infected mice models
  • Reduces virus-induced pathology in animal models
  • Exhibits a promising human pharmacokinetics profile
Catalog No. 50-003-73573 Supplier Medchemexpress LLC Supplier No. HY1531215MG
Leritrelvir (RAY1216) is an orally active SARS-CoV-2 main protease slow-tight inhibitor with a Ki of 8.6 nM. It is for research use only and not sold to patients.
  • Orally active SARS-CoV-2 main protease slow-tight inhibitor
  • Potent inhibitor with a Ki of 8.6 nM
  • Covalently attaches to the catalytic Cys145
  • Demonstrates antiviral activities against SARS-CoV-2 wild type ancestral strain and variants
  • Effectively prolongs survival in SARS-CoV-2 infected mice models
  • Reduces virus-induced pathology in animal models
  • Exhibits a promising human pharmacokinetics profile