Learn More
Medchemexpress LLC HY-112272A 5mg , Lerociclib (dihydrochloride) CAS:2097938-59-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112272A5MG
Encompass_Preferred
HY-112272A 5mg Lerociclib (dihydrochloride) CAS: 2097938-59-3 Lerociclib dihydrochloride (G1T38 dihydrochloride) is a potent and selective inhibitor of CDK4/CDK6, with IC50s of 1 nM and 2 nM for CDK4/CyclinD1 and CDK6/CyclinD3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1661