Learn More
Selleck Chemical LLC Lersivirine
SDP
Catalog No. 5000001018
Encompass_Preferred
Lersivirine (UK-453061) is a potent and selective inhibitor of nonnucleoside reverse transcriptase (NNRTI) with IC50 of 0 119 M
Catalog No. 50-000-01018 Supplier Selleck Chemical LLC Supplier No. S80555MG
Lersivirine (UK-453061) is a potent and selective inhibitor of nonnucleoside reverse transcriptase (NNRTI) with IC50 of 0 119 M