Learn More
Cayman Chemical Lersivirine, Purity: ≥98%, CAS: 473921-12-9, 50 mg
SDP
Catalog No. 502988701
Encompass_Preferred
An NNRTI (mean IC50 = 118 nM for the wild-type HIV-1 enzyme); inhibits mutant HIV-1 RTs with mean IC50 values within 10-fold of the wild-type enzyme; inhibits HIV-1 virus replication in MT-2 cells (EC50s = 5.04, 13.1, and 34.9 nM at MOIs of 0.005, 0.05, and 0.5, respectively); exhibits synergistic or additive effects when used in combination with a variety of antiretroviral agents
Catalog No. 50-298-8701 Supplier Cayman Chemical Supplier No. 2949750 MG
An NNRTI (mean IC50 = 118 nM for the wild-type HIV-1 enzyme); inhibits mutant HIV-1 RTs with mean IC50 values within 10-fold of the wild-type enzyme; inhibits HIV-1 virus replication in MT-2 cells (EC50s = 5.04, 13.1, and 34.9 nM at MOIs of 0.005, 0.05, and 0.5, respectively); exhibits synergistic or additive effects when used in combination with a variety of antiretroviral agents