Learn More
Selleck Chemical LLC Lesinurad S4640-20mg
SDP
Catalog No. 503108293
Encompass_Preferred
Lesinurad (RDEA-594) is a selective uric acid reabsorption inhibitor (SURI) under investigation for treatment of gout in combination with xanthine oxidase inhibitors Lesinurad is a URAT1 and OAT inhibitor is determined to be a substrate for the kidney transporters OAT1 and OAT3 with Km values of 0 85 and 2 M respectively
Catalog No. 50-310-8293 Supplier Selleck Chemical LLC Supplier No. S464020MG
Lesinurad (RDEA-594) is a selective uric acid reabsorption inhibitor (SURI) under investigation for treatment of gout in combination with xanthine oxidase inhibitors Lesinurad is a URAT1 and OAT inhibitor is determined to be a substrate for the kidney transporters OAT1 and OAT3 with Km values of 0 85 and 2 M respectively