Learn More
Selleck Chemical LLC Lesinurad sodium S5189-25mg
SDP
Catalog No. 503112190
Encompass_Preferred
Lesinurad (Selexipag RDEA-594) sodium (Zurampic) is a selective uric acid reabsorption inhibitor (SURI) under investigation for treatment of gout Lesinurad sodium inhibits the uric acid transporter URAT1 and Kidney transporter OAT with IC50/Km of 4 3 M/0 85 M and 3 5 M/2 M for OAT1 and OAT3 respectively
Catalog No. 50-311-2190 Supplier Selleck Chemical LLC Supplier No. S518925MG
Add to Cart
Add to Cart
Lesinurad (Selexipag RDEA-594) sodium (Zurampic) is a selective uric acid reabsorption inhibitor (SURI) under investigation for treatment of gout Lesinurad sodium inhibits the uric acid transporter URAT1 and Kidney transporter OAT with IC50/Km of 4 3 M/0 85 M and 3 5 M/2 M for OAT1 and OAT3 respectively