Learn More
Medchemexpress LLC Lesogaberan hydrochloride | 2925644-17-1 | 98.0% | 177.54 | C3H10ClFNO2P | 1 ML
SDP
Catalog No. 5000214636
Encompass_Preferred
Lesogaberan hydrochloride is a potent, selective GABAB receptor agonist provided as a ready-to-use 10 mM solution in DMSO for research applications. It is intended for use in receptor pharmacology, binding, and functional assays to probe GABAB-mediated signaling.
  • Potent and selective GABAB receptor agonist.
  • Supplied as a 10 mM solution in DMSO suitable for in vitro assays.
  • High purity, typically 98.0%, for reproducible results.
  • Molecular weight 177.54 g/mol; CAS number 2925644-17-1.
  • Suitable for receptor pharmacology, binding, and functional studies.
  • Available in multiple formats for experimental flexibility.
Catalog No. 50-002-14636 Supplier Medchemexpress LLC Supplier No. HY10061B10MM 1ML
Lesogaberan hydrochloride is a potent, selective GABAB receptor agonist provided as a ready-to-use 10 mM solution in DMSO for research applications. It is intended for use in receptor pharmacology, binding, and functional assays to probe GABAB-mediated signaling.
  • Potent and selective GABAB receptor agonist.
  • Supplied as a 10 mM solution in DMSO suitable for in vitro assays.
  • High purity, typically 98.0%, for reproducible results.
  • Molecular weight 177.54 g/mol; CAS number 2925644-17-1.
  • Suitable for receptor pharmacology, binding, and functional studies.
  • Available in multiple formats for experimental flexibility.