Learn More
Medchemexpress LLC Lesogaberan hydrochloride | 2925644-17-1 | 98.0% | 177.54 | C3H10ClFNO2P | 25 MG
SDP
Catalog No. 5000212839
Encompass_Preferred
Lesogaberan hydrochloride is a research-grade GABAB receptor agonist (hydrochloride salt) used as a pharmacological tool to study GABAB-mediated signaling and peripheral effects on lower esophageal sphincter relaxation. It is supplied as a solid reagent with supporting analytical documentation for laboratory research use only; not for human or veterinary use.
  • Potent, selective GABAB receptor agonist with EC50 of 8.6 nM.
  • High reported purity (98.0%).
  • Provided with datasheet, COA, and SDS for analytical verification.
  • Recommended storage: solid at 4°C under inert atmosphere to minimize degradation.
  • Suitable for in vitro pharmacology and biochemical assays.
Catalog No. 50-002-12839 Supplier Medchemexpress LLC Supplier No. HY10061B25MG
Lesogaberan hydrochloride is a research-grade GABAB receptor agonist (hydrochloride salt) used as a pharmacological tool to study GABAB-mediated signaling and peripheral effects on lower esophageal sphincter relaxation. It is supplied as a solid reagent with supporting analytical documentation for laboratory research use only; not for human or veterinary use.
  • Potent, selective GABAB receptor agonist with EC50 of 8.6 nM.
  • High reported purity (98.0%).
  • Provided with datasheet, COA, and SDS for analytical verification.
  • Recommended storage: solid at 4°C under inert atmosphere to minimize degradation.
  • Suitable for in vitro pharmacology and biochemical assays.