Learn More
TARGETMOL CHEMICALS INC LESOPITRON 5MG
SDP
Catalog No. 502580742
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lesopitron is an anxiolytic with pre- and postsynaptic 5-HT1A agonist activity and is more effective than 5-HT1A agonists in rat social interaction and marmoset anxiety models. lesopitron counteracts benzodiazepine withdrawal-induced anxiety in rodents with low acute toxicity and does not potentiate the effects of alcohol or barbiturates. purity: 98%
Catalog No. 50-258-0742 Supplier TARGETMOL CHEMICALS INC Supplier No. T25672
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lesopitron is an anxiolytic with pre- and postsynaptic 5-HT1A agonist activity and is more effective than 5-HT1A agonists in rat social interaction and marmoset anxiety models. lesopitron counteracts benzodiazepine withdrawal-induced anxiety in rodents with low acute toxicity and does not potentiate the effects of alcohol or barbiturates. purity: 98%