Learn More
Medchemexpress LLC Lesopitron dihydrochloride | 132449-89-9 | 98.0% | 393.74 g/mol | C15H23Cl3N6 | 5 MG
SDP
Catalog No. 5000245699
Encompass_Preferred
Lesopitron dihydrochloride is a selective, full agonist of the 5-HT1A receptor used in research to investigate serotonergic signaling and pharmacology. Supplier-reported data indicate activity in rodent tissues (IC50 ≈125 nM) and effects on adenylate cyclase activity and neuronal firing, supporting use in both in vitro and in vivo studies.
  • Selective full 5-HT1A receptor agonist.
  • Reported IC50 ≈125 nM in rat hippocampal membranes.
  • Modulates forskolin-stimulated adenylate cyclase activity.
  • Demonstrated activity in vitro and in vivo assays.
  • Supplied in small, research-oriented pack sizes for pharmacology studies.
Catalog No. 50-002-45699 Supplier Medchemexpress LLC Supplier No. HY1016095MG
Lesopitron dihydrochloride is a selective, full agonist of the 5-HT1A receptor used in research to investigate serotonergic signaling and pharmacology. Supplier-reported data indicate activity in rodent tissues (IC50 ≈125 nM) and effects on adenylate cyclase activity and neuronal firing, supporting use in both in vitro and in vivo studies.
  • Selective full 5-HT1A receptor agonist.
  • Reported IC50 ≈125 nM in rat hippocampal membranes.
  • Modulates forskolin-stimulated adenylate cyclase activity.
  • Demonstrated activity in vitro and in vivo assays.
  • Supplied in small, research-oriented pack sizes for pharmacology studies.