Learn More
TARGETMOL CHEMICALS INC LESOPITRON HCL 5MG
SDP
Catalog No. 502581509
Encompass_Preferred
Also available in 1 mg and bulk. Please contact Fisher for quotes. Lesopitron HCl is a full and selective 5-HT1A receptor agonist with IC50 of 125 nM in rat hippocampal membranes. purity: 98%
Catalog No. 50-258-1509 Supplier TARGETMOL CHEMICALS INC Supplier No. T11839L5MG
Also available in 1 mg and bulk. Please contact Fisher for quotes. Lesopitron HCl is a full and selective 5-HT1A receptor agonist with IC50 of 125 nM in rat hippocampal membranes. purity: 98%