Learn More
Cayman Chemical LestaurtInIb 10mg
SDP
Catalog No. 502951859
Encompass_Preferred
A staurosporine analog that potently inhibits JAK2 kinase (IC50 = 1 nM) and downstream targets STAT5 (IC50 = 10-30 nM) and STAT3 in a human erythroleukemic cell line expressing the JAK2V617F mutation; potently inhibits the epigenetic kinase PRK1 (PKN1) in vitro (IC50 = 8.6 nM)
Catalog No. 50-295-1859 Supplier Cayman Chemical Supplier No. 1209410 MG
A staurosporine analog that potently inhibits JAK2 kinase (IC50 = 1 nM) and downstream targets STAT5 (IC50 = 10-30 nM) and STAT3 in a human erythroleukemic cell line expressing the JAK2V617F mutation; potently inhibits the epigenetic kinase PRK1 (PKN1) in vitro (IC50 = 8.6 nM)