Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Let's Learn to Measure Activity Set
SCI_ED

Catalog No. S01293 Shop All Learning Resources, Inc Products
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01293 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01293 Supplier Hand2Mind Supplier No. 93401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides children with early experience in understanding volume and fractional relationships through a variety of activities and manipulatives.

  • Let's Learn to Measure Activity Set provides a rich experience in understanding volume and fractional relationships through a variety of activities and manipulatives.
  • Children will use the Rainbow Fraction Measuring Cups along with a Nestable Bucket Balance, and MathLink Cubes to learn about volume, weight, length, height, capacity, and more
  • These measuring cups all have the same radius so that they're equivalent proportions to their respective fraction sizes. This way kids can visually see the difference in the fraction sizes.
  • For over 50 years, hand2mind has encouraged hands-on learning and discovery, driving deeper understanding and helping children unlock their full potential. Hand2mind specializes in math manipulatives designed to help students learn by doing.
  • Particularly with fractions, our rainbow fraction tiles, rainbow fraction circles and rainbow fraction tower cubes make learning fractions as easy as learning the colors of the rainbow.
  • From early math to middle school math, hand2mind has you covered with products designed to enhance learning in the classroom and the world around us.
  • Through hands-on exploration and discovery we strive to find new ways to encourage students in math practice, literacy activities, STEM education and more!

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 2 to 4 Students
Product Type Fractions, Decimals, Percents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.