Learn More
Medchemexpress LLC HY-15233 5mg Medchemexpress, Letermovir CAS:917389-32-3 Purity:>98%
SDP
Catalog No. 502030593
Encompass_Preferred
Medchemexpress, HY-15233 5mg Letermovir CAS:917389-32-3 Letermovir is a novel inhibitor of CMV, which targets the viral terminase complex and remains active against virus resistant to DNA polymerase inhibitors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0593 Supplier Medchemexpress LLC Supplier No. HY152335MG
Medchemexpress, HY-15233 5mg Letermovir CAS:917389-32-3 Letermovir is a novel inhibitor of CMV, which targets the viral terminase complex and remains active against virus resistant to DNA polymerase inhibitors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.