Learn More
NEOGEN Letheen Agar Base Modified without Tween, 5 kg
SDP
Catalog No. 502015529
Encompass_Preferred
Letheen Agar Base Modified (without Tween) is used with Tween 80 (Polysorbate 80) for the isolation of microorganisms from cosmetics in a laboratory setting. Letheen Agar Base Modified is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Tween 80 (Polysorbate 80) to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds in testing of germicidal activity. Total neutralization of disinfectants is critical. Disinfectant residues can result in a false negative (no-growth) test.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-agar-base-modified-without-tween
Catalog No. 50-201-5529 Supplier NEOGEN Supplier No. 700003345
Letheen Agar Base Modified (without Tween) is used with Tween 80 (Polysorbate 80) for the isolation of microorganisms from cosmetics in a laboratory setting. Letheen Agar Base Modified is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Tween 80 (Polysorbate 80) to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds in testing of germicidal activity. Total neutralization of disinfectants is critical. Disinfectant residues can result in a false negative (no-growth) test.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-agar-base-modified-without-tween