Learn More
NEOGEN Letheen Broth Base Modified without Tween, 500 g
SDP
Catalog No. 502015525
Encompass_Preferred
Letheen Broth Base Modified without Tween is used with Polysorbate 80 for the recovery of microorganisms from cosmetics in a laboratory setting. Letheen Broth Base Modified without Tween is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Polysorbate 80 to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds in testing of germicidal activity. Total neutralization of disinfectants is critical. Disinfectant residues can result in a false negative (no-growth) test.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-base-modified-without-tween
Catalog No. 50-201-5525 Supplier NEOGEN Supplier No. 700003340
Letheen Broth Base Modified without Tween is used with Polysorbate 80 for the recovery of microorganisms from cosmetics in a laboratory setting. Letheen Broth Base Modified without Tween is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Polysorbate 80 to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds in testing of germicidal activity. Total neutralization of disinfectants is critical. Disinfectant residues can result in a false negative (no-growth) test.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-base-modified-without-tween