Learn More
NEOGEN Letheen Broth with Tween, Modified, 10 kg
SDP
Catalog No. 502015455
Encompass_Preferred
Letheen Broth with Tween, Modified is used for the recovery of microorganisms from cosmetics in a laboratory setting and is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Polysorbate 80 to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds in testing of germicidal activity. Total neutralization of disinfectants is critical as disinfectant residues can result in a false negative (no-growth) test result.Letheen Broth with Tween, Modified is based on the formula described in FDA Bacteriological Analytical Manual, and a modification of Letheen Broth Base.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-with-tween-modified
Catalog No. 50-201-5455 Supplier NEOGEN Supplier No. 700003265
Letheen Broth with Tween, Modified is used for the recovery of microorganisms from cosmetics in a laboratory setting and is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Polysorbate 80 to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds in testing of germicidal activity. Total neutralization of disinfectants is critical as disinfectant residues can result in a false negative (no-growth) test result.Letheen Broth with Tween, Modified is based on the formula described in FDA Bacteriological Analytical Manual, and a modification of Letheen Broth Base.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-with-tween-modified