Learn More
General Laboratory Products Letheen Broth, 500 mL, 12/cs
Catalog No. NC0916987
Encompass_Preferred
Letheen Broth - 500 ml, Easy-pour narrow mouth glass bottle, 12 per case. Base for Determining the Phenol Coefficient of Cationic Surface-Active Materials.
Catalog No. NC0916987 Supplier General Laboratory Products Supplier No. 500EZ1800500
Letheen Broth - 500 ml, Easy-pour narrow mouth glass bottle, 12 per case. Base for Determining the Phenol Coefficient of Cationic Surface-Active Materials.