Learn More
NEOGEN Letheen Broth Base without Tween, 5 kg
SDP
Catalog No. 502015571
Encompass_Preferred
Letheen Broth Base (no tween) is used with Tween 80 (Polysorbate 80) for the testing of samples containing quaternary ammonium compounds for antimicrobial activity in a laboratory setting. Letheen Broth Base (no tween) is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Polysorbate 80 to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds while testing germicidal activity. Letheen Agar is a modification of TGE Agar with the addition of Lecithin and Polysorbate 80.Letheen Broth Base was developed as a subculture medium for the neutralization of quaternary ammonium compounds in disinfectant testing.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-base-without-tween
Catalog No. 50-201-5571 Supplier NEOGEN Supplier No. 700003389
Letheen Broth Base (no tween) is used with Tween 80 (Polysorbate 80) for the testing of samples containing quaternary ammonium compounds for antimicrobial activity in a laboratory setting. Letheen Broth Base (no tween) is not intended for use in the diagnosis of disease or other conditions in humans.In 1948, Weber and Black described the value of a highly nutritional solid medium containing neutralizing agents for quaternary ammonium compounds in sanitizers. The addition of Lecithin and Polysorbate 80 to Tryptone Glucose Extract (TGE) Agar resulted in a medium that effectively neutralizes quaternary ammonium compounds while testing germicidal activity. Letheen Agar is a modification of TGE Agar with the addition of Lecithin and Polysorbate 80.Letheen Broth Base was developed as a subculture medium for the neutralization of quaternary ammonium compounds in disinfectant testing.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-base-without-tween