Learn More
MOLTOX Molecular Toxicology Inc Letheen Broth, 9 mL/tube, 10 tubes/box, Store in Refrigerator
SDP
Catalog No. 50948853
Encompass_Preferred
Used to determine the antimicrobial activity in quarternary ammonium compounds, primarily when evaluating disinfectants, germicides and cosmetics. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-853 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S134
Used to determine the antimicrobial activity in quarternary ammonium compounds, primarily when evaluating disinfectants, germicides and cosmetics. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.