Learn More
NEOGEN Letheen Broth with Tween, 10 kg
SDP
Catalog No. 502015488
Encompass_Preferred
Letheen Broth is primarily used for assessing the bactericidal activity of quaternary ammonium compounds, and is not intended for use in the diagnosis of disease or other conditions in humans.Letheen Broth is used to determine the phenol co-efficient of cationic surfactants as recommended by the Official Methods of Analysis of the Association of Official Analytical Chemists (AOAC). It is also used in hygiene swabbing protocols where it is necessary to neutralize quaternary ammonium compounds.A modification of FDA Broth, Letheen Broth contains lecithin to neutralize quaternary ammonium compounds and Polysorbate 80 to neutralize phenols, hexachlorophene, formalin and (with lecithin) ethanol.Letheen Broth is easily prepared and has a clear appearance aiding in visual inspection for growth.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-with-tween
Catalog No. 50-201-5488 Supplier NEOGEN Supplier No. 700003302
Letheen Broth is primarily used for assessing the bactericidal activity of quaternary ammonium compounds, and is not intended for use in the diagnosis of disease or other conditions in humans.Letheen Broth is used to determine the phenol co-efficient of cationic surfactants as recommended by the Official Methods of Analysis of the Association of Official Analytical Chemists (AOAC). It is also used in hygiene swabbing protocols where it is necessary to neutralize quaternary ammonium compounds.A modification of FDA Broth, Letheen Broth contains lecithin to neutralize quaternary ammonium compounds and Polysorbate 80 to neutralize phenols, hexachlorophene, formalin and (with lecithin) ethanol.Letheen Broth is easily prepared and has a clear appearance aiding in visual inspection for growth.For more information visit https://www.neogen.com/solutions/Microbiology/letheen-broth-with-tween