Learn More
Aviva Systems Biology Corp LETM1 Antibody - middle region (ARP46881_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46881_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LETM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LETM1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59427