Learn More
Medchemexpress LLC Letosteine | 53943-88-7 | 99.8% | 279.38 g·mol⁻1 | C10H17NO4S2 | 200 MG
SDP
Catalog No. 5000241368
Encompass_Preferred
Letosteine is an orally active mucolytic expectorant used in research on acute and chronic respiratory diseases. It scavenges reactive oxygen species, dissolves bronchial mucus, reduces respiratory inflammation, and helps restore gas exchange and natural lung defenses.
  • Scavenges reactive oxygen species to reduce oxidative stress.
  • Dissolves bronchial mucus to improve airway clearance.
  • Reduces respiratory inflammation in experimental models.
  • Suitable for preclinical research on bronchopneumopathies and respiratory disorders.
  • Available in multiple research pack sizes for experimental flexibility.
Catalog No. 50-002-41368 Supplier Medchemexpress LLC Supplier No. HY107355200MG
Letosteine is an orally active mucolytic expectorant used in research on acute and chronic respiratory diseases. It scavenges reactive oxygen species, dissolves bronchial mucus, reduces respiratory inflammation, and helps restore gas exchange and natural lung defenses.
  • Scavenges reactive oxygen species to reduce oxidative stress.
  • Dissolves bronchial mucus to improve airway clearance.
  • Reduces respiratory inflammation in experimental models.
  • Suitable for preclinical research on bronchopneumopathies and respiratory disorders.
  • Available in multiple research pack sizes for experimental flexibility.