Learn More
TARGETMOL CHEMICALS INC LETOSTEINE 5MG
SDP
Catalog No. 502579612
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Letosteine (Viscotiol) is an orally active effective and safe expectorant that can be used in the study of acute or chronic respiratory diseases for example bronchopulmonary disease. Letosteine dissolves bronchial mucus and reduces symptoms of respiratory inflammation and restores gas exchanges and natural defense mechanisms in the lung[1] [2] [3]. purity: 96%
Catalog No. 50-257-9612 Supplier TARGETMOL CHEMICALS INC Supplier No. T605225MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Letosteine (Viscotiol) is an orally active effective and safe expectorant that can be used in the study of acute or chronic respiratory diseases for example bronchopulmonary disease. Letosteine dissolves bronchial mucus and reduces symptoms of respiratory inflammation and restores gas exchanges and natural defense mechanisms in the lung[1] [2] [3]. purity: 96%