Learn More
Medchemexpress LLC HY-14248 100mg , Letrozole CAS:112809-51-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14248100MG
Encompass_Preferred
HY-14248 100mg Letrozole CAS: 112809-51-5 Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0852