Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Letrozole

Catalog No. AC463190010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463190010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463190010 Supplier Thermo Scientific Chemicals Supplier No. 463190010

CAS: 112809-51-5 | C17H11N5 | 285.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112809-51-5
Molecular Formula C17H11N5
Molecular Weight (g/mol) 285.31
InChI Key HPJKCIUCZWXJDR-UHFFFAOYSA-N
IUPAC Name 4-[(4-cyanophenyl)(1H-1,2,4-triazol-1-yl)methyl]benzonitrile
SMILES N#CC1=CC=C(C=C1)C(N1C=NC=N1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 185.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very slightly soluble in ethanol, slightly soluble in methanol, soluble in acetonitrile
Formula Weight 285.3
Chemical Name or Material Letrozole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Causes skin irritation.

Causes serious eye irritation.

Harmful if inhaled.

May damage fertility.

May damage the unborn child.

May cause ha
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

Avoid contact during pregnancy and while nursing

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.