Learn More
Cayman Chemical Letrozole-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1133712-96-5, 1 mg
SDP
Catalog No. 502933468
Encompass_Preferred
An internal standard for the quantification of letrozole by GC- or LC-MS
Catalog No. 50-293-3468 Supplier Cayman Chemical Supplier No. 265171 MG
An internal standard for the quantification of letrozole by GC- or LC-MS