Learn More
Cayman Chemical Letrozole-d4 500ug
SDP
Catalog No. 502934572
Encompass_Preferred
An internal standard for the quantification of letrozole by GC- or LC-MS
Catalog No. 50-293-4572 Supplier Cayman Chemical Supplier No. 26517500 UG
An internal standard for the quantification of letrozole by GC- or LC-MS