Learn More
Medchemexpress LLC Letrozole (Standard) | 112809-51-5 | C17H11N5 | 200 MG
SDP
Catalog No. 5000353575
Encompass_Preferred
Letrozole (CGS 20267) is a potent, selective, reversible, and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. It selectively inhibits estrogen biosynthesis and can be used for breast cancer research.
  • Potent, selective, reversible, and orally active non-steroidal inhibitor of aromatase
  • Selectively inhibits estrogen biosynthesis
  • Used for breast cancer research
  • Exhibits anti-tumor effects in rats
  • Inhibits growth of MCF-7 epithelial breast cancer cells
Catalog No. 50-003-53575 Supplier Medchemexpress LLC Supplier No. HY14248200MG
Letrozole (CGS 20267) is a potent, selective, reversible, and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. It selectively inhibits estrogen biosynthesis and can be used for breast cancer research.
  • Potent, selective, reversible, and orally active non-steroidal inhibitor of aromatase
  • Selectively inhibits estrogen biosynthesis
  • Used for breast cancer research
  • Exhibits anti-tumor effects in rats
  • Inhibits growth of MCF-7 epithelial breast cancer cells