Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lets Tackle Math!™
SCI_ED

Catalog No. S78999
Change view
Click to view available options
Let's Tackle Math
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S78999 Let's Tackle Math
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S78999 Supplier Learning Resources, Inc Supplier No. LER 0223

missing translation for 'validMainframeMsg'

Tackle math with critters!

Make sorting and counting practice fun. This 252-piece counter set features assorted colorful bugs, worms and frogs to sort by color, shape and size. Also includes a 25-page stand-up activity book filled with 48 engaging, self-checking activities, sorting tray and sturdy tackle box.

Specifications

Color Assorted
Description Lets Tackle Math!™ Counter Set
For Use With (Application) Sorting and counting practice
Includes 252-piece set of assorted colorful bugs, worms and frogs to sort by color, shape and size; 25-page stand-up activity book filled with 48 engaging, self-checking activities; Sorting tray; Sturdy tackle box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.