Learn More
Grainger Letter Label Character S PK10
Catalog No. 502750511
Encompass_Preferred
The Brady Letter Label is a 2 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They have high adhesion and are resistant to most solvents and weather conditions. The labels come in a pack of 250 and have a temperature range of -40°F to 176°F. Overall, these versatile vinyl letter labels are ideal for creating clear identifiers in industrial environments.
Catalog No. 50-275-0511 Supplier Grainger Supplier No. 14R128
The Brady Letter Label is a 2 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They have high adhesion and are resistant to most solvents and weather conditions. The labels come in a pack of 250 and have a temperature range of -40°F to 176°F. Overall, these versatile vinyl letter labels are ideal for creating clear identifiers in industrial environments.