Learn More
Medchemexpress LLC HY-108900 1mg Medchemexpress, Leu-AMS CAS:288591-93-5 Purity:>98%
SDP
Catalog No. 501968632
Encompass_Preferred
Medchemexpress, HY-108900 1mg Leu-AMS CAS:288591-93-5 Leu-AMS (compound 6), a leucine analogue, is a potent inhibitor of leucyl-tRNA synthetase (LRS) with an IC50 of 22.34 nM, which inhibits the catalytic activity of LRS but did not affect the leucine-induced mTORC1 activation. Leu-AMS shows cytotoxicity in cancer cells and normal cells, and inhibits the growth of bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8632 Supplier Medchemexpress LLC Supplier No. HY1089001MG
Medchemexpress, HY-108900 1mg Leu-AMS CAS:288591-93-5 Leu-AMS (compound 6), a leucine analogue, is a potent inhibitor of leucyl-tRNA synthetase (LRS) with an IC50 of 22.34 nM, which inhibits the catalytic activity of LRS but did not affect the leucine-induced mTORC1 activation. Leu-AMS shows cytotoxicity in cancer cells and normal cells, and inhibits the growth of bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.