Learn More
Medchemexpress LLC Leu-AMS R enantiomer | 459.48 | 10 MG
SDP
Catalog No. 501932334
Encompass_Preferred
Leu-AMS R enantiomer is the R enantiomer of Leu-AMS. Leu-AMS is a potent inhibitor of leucyl-tRNA synthetase (LRS) and inhibits the growth of bacteria. For research use only.
  • Potent inhibitor of leucyl-tRNA synthetase (LRS).
  • Inhibits the growth of bacteria.
Catalog No. 50-193-2334 Supplier Medchemexpress LLC Supplier No. HY108900A1MG
Leu-AMS R enantiomer is the R enantiomer of Leu-AMS. Leu-AMS is a potent inhibitor of leucyl-tRNA synthetase (LRS) and inhibits the growth of bacteria. For research use only.
  • Potent inhibitor of leucyl-tRNA synthetase (LRS).
  • Inhibits the growth of bacteria.