Learn More
Enzo Life Sciences Leu-pNA (100mg). CAS: 4178-93-2
SDP
Supplier:  Enzo Life Sciences BMLP2300100
Encompass_Preferred
Substrate for aminopeptidases such as aminopeptidase N (APN) cystinyl aminopeptidase, and leucine aminopeptidase (LAP). This substrate can be used for in vitro assays with purified enzyme, or with cell extracts. Also available: calibration standard pNA (BML-KI145). Alternative name: Aminopeptidase substrate (chromogenic). Purity: ≥99% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-201-1298