Learn More
Carbosynth LLC. Leu-pNA, 1g, MW: 251.28, Molecular formula: C12H17N3O3, CAS# 4178-93-2
SDP
Supplier:  Carbosynth LLC. SLN30141G
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: L-Leucine p-Nitroanilide, Purity: , Grade AA, Application: Substrate for Aminopeptidase, Storage Temperature: -20C, References: G. Peleiderer, In, Proteolytic Enzymes, Methods in Enzymology, Vol. 19, (G.E. Perlmann and L. Lorand, Eds.), Academic Press, New York, 1970, pp. 514-521.
Catalog No. 50-168-7122