Learn More
Bachem (Leu15)-Gastrin I (human)
SDP
Catalog No. 50259697
Encompass_Preferred
(Leu15)-Gastrin I (human), Pyr-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Leu-Asp-Phe-NH2, 39024-57-2, C98H126N20O31, Mr 2080.20, 1mg. (Leu15)-Gastrin-17 I (human), (Pyr18,Leu22,Leu32)-Gastrin I (18-34) (porcine). This Gastrin I analog is as active as the native sequence, but more stable. The activity loss of Gastrin due to Met sulfoxide formation is avoided.
Catalog No. 50-259-697 Supplier Bachem Supplier No. 4011661.0001
Add to Cart
Add to Cart
(Leu15)-Gastrin I (human), Pyr-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Leu-Asp-Phe-NH2, 39024-57-2, C98H126N20O31, Mr 2080.20, 1mg. (Leu15)-Gastrin-17 I (human), (Pyr18,Leu22,Leu32)-Gastrin I (18-34) (porcine). This Gastrin I analog is as active as the native sequence, but more stable. The activity loss of Gastrin due to Met sulfoxide formation is avoided.