Learn More
AnaSpec [Leu27]-Melan-A, MART 1 (26 - 35), 1mg
Supplier:  AnaSpec AS61013
Encompass
Sequence: ELAGIGILTV, Molecular Weight 985.8, This Melan-A (26-35) analog, Leu substituted for Ala at position 27, shows better HLA-A*0201 binding properties as well as better immunogenicity and antigenicity than the natural Melan-A (26-35).
Catalog No. NC0830659