Learn More
Bachem (Leu31,Pro34)-Neuropeptide Y (human, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4031098.1000
Encompass_Preferred
(Leu31,Pro34)-Neuropeptide Y (human, rat) Trifluoroacetate, H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Leu-Thr-Arg-Pro-Arg-Tyr-NH2, 132699-73-1, C189H284N54O56S, Mr 4240.73, 1mg. Specific Y1 receptor agonist.
Catalog No. 50-259-735