Learn More
CPC Scientific H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Leu-Thr-Arg-Pro-Arg-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP101B
Encompass
ONE-LETTER SEQUENCE: YPSKPDNPGEDAPAEDMARYYSALRHYINLLTRPRY-NH2MOLECULAR FORMULA: C189H284N54O56S1MOLECULAR WEIGHT:4240.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [132699-73-1], RESEARCH AREA: NeuropeptidesREFERENCES: Futilendorf, I. et al. Proc. Natl. Acad. Sci. USA 87, 182 (1990).; Labelle, M. et al. Peptides 18, 801 (1997).; Khiat, AJ. Pept. Res. 51, 317 (1998).; Chen, S. and RJ. Cheung, Biomed. Sci. 11, 781 (2004).; Chen, S. and RJ. Cheung, Biomed. Sci.12, 267 (2005).
Catalog No. 50-210-9926