Learn More
Medchemexpress LLC HY-P0288 10mg Medchemexpress, [Leu5]-Enkephalin CAS:58822-25-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP028810MG
Encompass_Preferred
Medchemexpress, HY-P0288 10mg [Leu5]-Enkephalin CAS:58822-25-6 [Leu5]-Enkephalin is a pentapeptides with morphine like properties. [Leu5]-Enkephalin is a five amino acid endogenous peptide that acts as an agonist at opioid receptors . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4099