Learn More
CPC Scientific H-Asp-Arg-Val-Tyr-Ile-His-Pro-Leu-His-Leu-Leu-Tyr-Tyr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110364
Encompass
ONE-LETTER SEQUENCE: DRVYIHPLHLLYYSMOLECULAR FORMULA: C86H125N21O21MOLECULAR WEIGHT:1789.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-211-0364 Supplier CPC Scientific Supplier No. RENI008A
ONE-LETTER SEQUENCE: DRVYIHPLHLLYYSMOLECULAR FORMULA: C86H125N21O21MOLECULAR WEIGHT:1789.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular