Learn More
Medchemexpress LLC Leucic acid (α-Hydroxyisocaproic acid) | 498-36-2 | 99.2% | 132.16 | 500 MG
SDP
Catalog No. 5000403994
Encompass_Preferred
Leucic acid (α-Hydroxyisocaproic acid) is an orally active end-product of leucine's microbial metabolism. It binds to HCAR2, altering AMPK and ERK1/2 phosphorylation. It suppresses lipid synthesis, promotes catabolism, reduces adiposity, enhances lean mass and exercise capacity, attenuates myotube atrophy. It also suppresses pro-inflammatory cytokine secretion and inflammation-related gene mRNA expression. Useful for obesity research.
  • Orally active end-product of leucine metabolism
  • Suppresses lipid synthesis, promotes catabolism, reduces adiposity
  • Enhances lean mass and exercise capacity
  • Suppresses pro-inflammatory cytokine secretion and inflammation
  • Decreases basal protein synthesis; attenuates myotube atrophy
  • Activates AMPK and p38MAPK signaling
  • Useful for obesity research
Catalog No. 50-004-03994 Supplier Medchemexpress LLC Supplier No. HY30216A500MG
Leucic acid (α-Hydroxyisocaproic acid) is an orally active end-product of leucine's microbial metabolism. It binds to HCAR2, altering AMPK and ERK1/2 phosphorylation. It suppresses lipid synthesis, promotes catabolism, reduces adiposity, enhances lean mass and exercise capacity, attenuates myotube atrophy. It also suppresses pro-inflammatory cytokine secretion and inflammation-related gene mRNA expression. Useful for obesity research.
  • Orally active end-product of leucine metabolism
  • Suppresses lipid synthesis, promotes catabolism, reduces adiposity
  • Enhances lean mass and exercise capacity
  • Suppresses pro-inflammatory cytokine secretion and inflammation
  • Decreases basal protein synthesis; attenuates myotube atrophy
  • Activates AMPK and p38MAPK signaling
  • Useful for obesity research