Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leucic acid (standard) | 498-36-2 | 98.6% | 132.16 | 25 MG
SDP

Catalog No. 5000402268
Encompass_Preferred

Leucic acid is supplied as a Standard, indicating its suitability for use as a reference material. It has an assay purity of 98.64% determined by Q-NMR. The product is white to off-white solid and is stored in a brown glass bottle at -20°C, sealed and dry, away from light and moisture. Its traceability is fully qualified and calibrated to ISO 17025 requirements, with all calibrations utilizing institute of metrology traceable weights, externally calibrated by an ISO 17025 accredited calibration laboratory to metrology institute standards.

  • Suitable for use as a reference material
  • Assay purity of 98.64% determined by Q-NMR
  • White to off-white solid appearance
  • Stored in a brown glass bottle at -20°C, sealed and dry
  • Fully qualified and calibrated to ISO 17025 requirements
  • Calibrations utilize metrology institute traceable weights

Catalog No. 50-004-02268 Supplier Medchemexpress LLC Supplier No. 25MGHY30216AR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Leucic acid is supplied as a Standard, indicating its suitability for use as a reference material. It has an assay purity of 98.64% determined by Q-NMR. The product is white to off-white solid and is stored in a brown glass bottle at -20°C, sealed and dry, away from light and moisture. Its traceability is fully qualified and calibrated to ISO 17025 requirements, with all calibrations utilizing institute of metrology traceable weights, externally calibrated by an ISO 17025 accredited calibration laboratory to metrology institute standards.

  • Suitable for use as a reference material
  • Assay purity of 98.64% determined by Q-NMR
  • White to off-white solid appearance
  • Stored in a brown glass bottle at -20°C, sealed and dry
  • Fully qualified and calibrated to ISO 17025 requirements
  • Calibrations utilize metrology institute traceable weights

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.