Learn More
Selleck Chemical LLC Leucic acid
SDP
Catalog No. 5000003003
Encompass_Preferred
Leucic acid ((S)- -hydroxyisocaproic acid) is a metabolite of the branched chain amino acid known as leucine which plays a significant role in the synthesis of muscle protein
Catalog No. 50-000-03003 Supplier Selleck Chemical LLC Supplier No. S604625MG
Leucic acid ((S)- -hydroxyisocaproic acid) is a metabolite of the branched chain amino acid known as leucine which plays a significant role in the synthesis of muscle protein