Learn More
Medchemexpress LLC (±)-Leucine-d10 | 29909-01-1 | 98.0% | 5 MG
SDP
Catalog No. 5000412745
Encompass_Preferred
(±)-Leucine-d10 is a deuterium-labeled form of (±)-Leucine. (±)-Leucine (DL-Leucine) is an isomer of Leucine, and it acts as a chemosterilant and dietary additive. It has been shown to inhibit the growth of *Escherichia coli* HfrH by 92.08%. Stable heavy isotopes of hydrogen, carbon, and other elements, when incorporated into drug molecules, are largely used as tracers for quantitation during drug development. Deuteration, specifically, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12745 Supplier Medchemexpress LLC Supplier No. HYB1674S5MG
(±)-Leucine-d10 is a deuterium-labeled form of (±)-Leucine. (±)-Leucine (DL-Leucine) is an isomer of Leucine, and it acts as a chemosterilant and dietary additive. It has been shown to inhibit the growth of *Escherichia coli* HfrH by 92.08%. Stable heavy isotopes of hydrogen, carbon, and other elements, when incorporated into drug molecules, are largely used as tracers for quantitation during drug development. Deuteration, specifically, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.