Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leucine dehydrogenase, Microorganism | 9082-71-7 | ≥25 U/mg protein | 43 kDa | 200 U
SDP

Catalog No. 5000400387
Encompass_Preferred

Leucine dehydrogenase, Microorganism (EC 1.4.1.9) is an enzyme purified from Bacillus spheroides. It catalyzes the oxidative deamination of various L-amino acids, including L-leucine, L-valine, L-isoleucine, L-norvaline, L-alpha-aminobutyrate, and L-norleucine. This enzyme also facilitates the reductive amination of their corresponding keto analogues.

  • Isoelectric point: 6.6
  • Km value: 2.6×10⁻⁴ M (NAD), 2.0×10⁻³ M (L-leucine), 6.8×10⁻⁴ M (α-ketoisocaproate), 4.2×10⁻² M (NH₄Cl), 2.3×10⁻⁴ M (NADH)
  • pH stability: 6.0-11.0 (25°C, 15 hr)
  • Optimum temperature: 55-60°C
  • Inhibitor: Hg²+
  • For research use only
  • Can be dissolved in water (1 mg/mL) or in buffer according to specific experimental references

Catalog No. 50-004-00387 Supplier Medchemexpress LLC Supplier No. HYP2768200U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leucine dehydrogenase, Microorganism (EC 1.4.1.9) is an enzyme purified from Bacillus spheroides. It catalyzes the oxidative deamination of various L-amino acids, including L-leucine, L-valine, L-isoleucine, L-norvaline, L-alpha-aminobutyrate, and L-norleucine. This enzyme also facilitates the reductive amination of their corresponding keto analogues.

  • Isoelectric point: 6.6
  • Km value: 2.6×10⁻⁴ M (NAD), 2.0×10⁻³ M (L-leucine), 6.8×10⁻⁴ M (α-ketoisocaproate), 4.2×10⁻² M (NH₄Cl), 2.3×10⁻⁴ M (NADH)
  • pH stability: 6.0-11.0 (25°C, 15 hr)
  • Optimum temperature: 55-60°C
  • Inhibitor: Hg²+
  • For research use only
  • Can be dissolved in water (1 mg/mL) or in buffer according to specific experimental references

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.