Learn More
Medchemexpress LLC (±)-Leucine (DL-Leucine) | 328-39-2 | 99.95% | 100 G
SDP
Catalog No. 5000413134
Encompass_Preferred
(±)-Leucine, also known as DL-Leucine, is an isomer of the amino acid Leucine. It functions as both a chemosterilant and a dietary additive. Research indicates that this compound is effective in inhibiting bacterial growth, specifically demonstrating a 92.08% inhibition of Escherichia coli HfrH. Additionally, (±)-Leucine has been observed to enhance the average catch in certain applications by increasing the quantity of pheromone or the frequency of its production.
  • Isomer of Leucine, also known as DL-Leucine or (RS)-Leucine.
  • Functions as a chemosterilant and dietary additive.
  • Demonstrates significant inhibitory activity against Escherichia coli HfrH (92.08% inhibition).
  • Reported to increase average catch by influencing pheromone production.
  • High purity of 99.95%.
  • Comprehensive documentation, including Data Sheet and SDS, is available.
Catalog No. 50-004-13134 Supplier Medchemexpress LLC Supplier No. HYB1674100G
(±)-Leucine, also known as DL-Leucine, is an isomer of the amino acid Leucine. It functions as both a chemosterilant and a dietary additive. Research indicates that this compound is effective in inhibiting bacterial growth, specifically demonstrating a 92.08% inhibition of Escherichia coli HfrH. Additionally, (±)-Leucine has been observed to enhance the average catch in certain applications by increasing the quantity of pheromone or the frequency of its production.
  • Isomer of Leucine, also known as DL-Leucine or (RS)-Leucine.
  • Functions as a chemosterilant and dietary additive.
  • Demonstrates significant inhibitory activity against Escherichia coli HfrH (92.08% inhibition).
  • Reported to increase average catch by influencing pheromone production.
  • High purity of 99.95%.
  • Comprehensive documentation, including Data Sheet and SDS, is available.